1-Phenethylguanidine sulfate

Supplier: Matrix Scientific

066684-250MG
101932-132EA 280 USD
101932-132
1-Phenethylguanidine sulfate
1-Phenethylguanidine sulfate

Hazard: Irritant

Formula: C₉H₁₅N₃O₄S
MW: 261.3 g/mol
MDL Number: MFCD00035581
CAS Number: 202401-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR