8-Bromo-5-chloro-2-(trifluoromethyl)-1,6-naphthyridine
Supplier: Matrix Scientific
066683-1G
101932-130EA
516.92
USD
101932-130
8-Bromo-5-chloro-2-(trifluoromethyl)-1,6-naphthyridine
8-Bromo-5-chloro-2-(trifluoromethyl)-1,6-naphthyridine
Hazard: Irritant
Formula:
C₉H₃BrClF₃N₂ |
MDL Number:
MFCD08567942 CAS Number: 890301-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...