Ethyl-2-(3-iodophenoxy)acetate
Supplier: Matrix Scientific
066680-1G
101932-124EA
481.54
USD
101932-124
Ethyl-2-(3-iodophenoxy)acetate
Ethyl-2-(3-iodophenoxy)acetate
Hazard: Irritant
Formula:
C₁₀H₁₁IO₃ MW: 306.1 g/mol |
MDL Number:
MFCD06659701 CAS Number: 90888-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...