2-Cyclopropylbenzonitrile

Supplier: Matrix Scientific

066679-500MG
101932-122EA 380 USD
101932-122
2-Cyclopropylbenzonitrile
2-Cyclopropylbenzonitrile

Hazard: Irritant

Formula: C₁₀H₁₀N
MW: 143.19 g/mol
MDL Number: MFCD00463731
CAS Number: 3154-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR