Ethyl-2-m-tolylpyrimidine-5-carboxylate

Supplier: Matrix Scientific

066678-500MG
101932-120EA 573.85 USD
101932-120
Ethyl-2-m-tolylpyrimidine-5-carboxylate
Ethyl-2-m-tolylpyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MW: 242.28 g/mol
MDL Number: MFCD09863220
CAS Number: 954227-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR