1-Boc-4-[4-(methylsulfonyl)-2-nitrophenyl]piperidin-4-amine

Supplier: Matrix Scientific

066659-1G
101932-080EA 481.54 USD
101932-080
1-Boc-4-[4-(methylsulfonyl)-2-nitrophenyl]piperidin-4-amine
1-Boc-4-[4-(methylsulfonyl)-2-nitrophenyl]piperidin-4-amine

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₆S
MDL Number: MFCD06245536
CAS Number: 849035-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR