1-[2-Chloro-4-methyl-5-(methylsulfonyl)phenyl]piperazine

Supplier: Matrix Scientific

066658-1G
101932-078EA 369.23 USD
101932-078
1-[2-Chloro-4-methyl-5-(methylsulfonyl)phenyl]piperazine
1-[2-Chloro-4-methyl-5-(methylsulfonyl)phenyl]piperazine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂S
MDL Number: MFCD04037928
CAS Number: 849035-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR