2-Amino-6-bromobenzonitrile ≥98%

Supplier: Matrix Scientific

066622-1G 066622-5G
101931-996EA 75.38 USD
101931-996 101931-994
2-Amino-6-bromobenzonitrile ≥98%
2-Amino-6-bromobenzonitrile

Hazard: Toxic

Formula: C₇H₅BrN₂
MW: 197.02 g/mol
MDL Number: MFCD09834769
CAS Number: 77326-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR