2-Amino-4-bromobenzonitrile ≥98%

Supplier: Matrix Scientific

066621-5G 066621-25G
101931-992EA 72.31 USD
101931-992 101931-990
2-Amino-4-bromobenzonitrile ≥98%
2-Amino-4-bromobenzonitrile

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
Boiling Pt: 317.8 °C
Melting Pt: 162…163 °C
Density: 1.69
Flash Pt: 146 °C
MDL Number: MFCD08752638
CAS Number: 304858-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR