2-(2,4-Dichlorophenyl)-1H-benzimidazole

Supplier: Matrix Scientific

066592-500MG
101931-962EA 243.08 USD
101931-962
2-(2,4-Dichlorophenyl)-1H-benzimidazole
2-(2,4-Dichlorophenyl)-1H-benzimidazole

Hazard: Irritant

Formula: C₁₃H₈Cl₂N₂
MDL Number: MFCD00224350
CAS Number: 14225-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR