1-Isopropyl-1H-benzimidazole-2-sulfonic acid

Supplier: Matrix Scientific

066578-500MG
101931-934EA 243.08 USD
101931-934
1-Isopropyl-1H-benzimidazole-2-sulfonic acid
1-Isopropyl-1H-benzimidazole-2-sulfonic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃S
MDL Number: MFCD02630562
CAS Number: 378764-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR