1-Isobutyl-1H-benzimidazole-2-thiol
Supplier: Matrix Scientific
066572-500MG
101931-922EA
183.08
USD
101931-922
1-Isobutyl-1H-benzimidazole-2-thiol
1-Isobutyl-1H-benzimidazole-2-thiol
Hazard: Irritant
Formula:
C₁₁H₁₄N₂S |
MDL Number:
MFCD02074359 CAS Number: 55489-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...