(1-Ethyl-1H-imidazol-2-yl)methanol

Supplier: Matrix Scientific

066568-500MG
101931-914EA 364.62 USD
101931-914
(1-Ethyl-1H-imidazol-2-yl)methanol
(1-Ethyl-1H-imidazol-2-yl)methanol

Hazard: Irritant

Formula: C₆H₁₀N₂O
MDL Number: MFCD09881565
CAS Number: 63634-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR