2-(3-Hydroxypropyl)benzimidazole

Supplier: Matrix Scientific

Synonyms: 3-(2-Benzimidazolyl)propanol2-Benzimidazolepropanol3-(1H-Benzo[d]imidazol-2-yl)propan-1-ol

066564-500MG
101931-906EA 146.15 USD
101931-906
2-(3-Hydroxypropyl)benzimidazole
2-(3-Hydroxypropyl)benzimidazole

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
MDL Number: MFCD00022688
CAS Number: 2403-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR