(2Z)-2-(1,3-Benzodioxol-5-ylmethylene)-6-hydroxy-1-benzofuran-3(2H)-one

Supplier: Matrix Scientific

066549-500MG
101931-876EA 484.62 USD
101931-876
(2Z)-2-(1,3-Benzodioxol-5-ylmethylene)-6-hydroxy-1-benzofuran-3(2H)-one
(2Z)-2-(1,3-Benzodioxol-5-ylmethylene)-6-hydroxy-1-benzofuran-3(2H)-one

Hazard: Irritant

Formula: C₁₆H₁₀O₅
MDL Number: MFCD06493721
CAS Number: 32396-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR