(2Z)-2-(4-Fluorobenzylidene)-6-hydroxy-1-benzofuran-3(2H)-one

Supplier: Matrix Scientific

066525-500MG
101931-828EA 484.62 USD
101931-828
(2Z)-2-(4-Fluorobenzylidene)-6-hydroxy-1-benzofuran-3(2H)-one
(2Z)-2-(4-Fluorobenzylidene)-6-hydroxy-1-benzofuran-3(2H)-one

Hazard: Irritant

Formula: C₁₅H₉FO₃
MDL Number: MFCD04065085
CAS Number: 139311-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR