7-Hydroxy-3-(2-methoxyphenyl)-2-methyl-4H-chromen-4-one

Supplier: Matrix Scientific

066512-500MG
101931-802EA 364.62 USD
101931-802
7-Hydroxy-3-(2-methoxyphenyl)-2-methyl-4H-chromen-4-one
7-Hydroxy-3-(2-methoxyphenyl)-2-methyl-4H-chromen-4-one

Hazard: Irritant

Formula: C₁₇H₁₄O₄
MDL Number: MFCD00205543
CAS Number: 308087-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR