1-(2-Methoxy-5-methylphenyl)-1H-pyrrole-2,5-dione

Supplier: Matrix Scientific

066489-500MG
101931-756EA 243.08 USD
101931-756
1-(2-Methoxy-5-methylphenyl)-1H-pyrrole-2,5-dione
1-(2-Methoxy-5-methylphenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD00030663
CAS Number: 199464-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR