N-(5-Chloro-2-methoxyphenyl)-N'-(4,6-dimethylpyrimidin-2-yl)guanidine

Supplier: Matrix Scientific

066419-500MG
101931-616EA 183.08 USD
101931-616
N-(5-Chloro-2-methoxyphenyl)-N'-(4,6-dimethylpyrimidin-2-yl)guanidine
N-(5-Chloro-2-methoxyphenyl)-N'-(4,6-dimethylpyrimidin-2-yl)guanidine

Hazard: Irritant

Formula: C₁₄H₁₆ClN₅O
MDL Number: MFCD02722938
CAS Number: 377061-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR