(4-Phenylpyrimidin-2-yl)cyanamide
Supplier: Matrix Scientific
066365-500MG
101931-508EA
243.08
USD
101931-508
(4-Phenylpyrimidin-2-yl)cyanamide
(4-Phenylpyrimidin-2-yl)cyanamide
Hazard: Irritant
Formula:
C₁₁H₈N₄ |
MDL Number:
MFCD19103635 CAS Number: 140369-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...