7-Amino-2-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carbonitrile

Supplier: Matrix Scientific

066354-500MG
101931-486EA 484.62 USD
101931-486
7-Amino-2-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carbonitrile
7-Amino-2-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carbonitrile

Hazard: Irritant

Formula: C₇H₆N₆
MDL Number: MFCD08088404
CAS Number: 896666-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR