4-(2-Methoxyphenyl)-1,4-dihydro[1,3,5]triazino[1,2-a]benzimidazol-2-amine

Supplier: Matrix Scientific

066292-500MG
101931-362EA 364.62 USD
101931-362
4-(2-Methoxyphenyl)-1,4-dihydro[1,3,5]triazino[1,2-a]benzimidazol-2-amine
4-(2-Methoxyphenyl)-1,4-dihydro[1,3,5]triazino[1,2-a]benzimidazol-2-amine

Hazard: Irritant

Formula: C₁₆H₁₅N₅O
MDL Number: MFCD04159896
CAS Number: 669718-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR