1-(7-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)ethanone

Supplier: Matrix Scientific

066282-500MG
101931-342EA 146.15 USD
101931-342
1-(7-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)ethanone
1-(7-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)ethanone

Hazard: Irritant

Formula: C₈H₈N₄O
MDL Number: MFCD00244648
CAS Number: 320416-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR