4,5,6,7-Tetrahydro-1H-indazole

Supplier: Matrix Scientific

066216-500MG
101931-210EA 552.31 USD
101931-210
4,5,6,7-Tetrahydro-1H-indazole
4,5,6,7-Tetrahydro-1H-indazole

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.17
MDL Number: MFCD00014565
CAS Number: 2305-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR