N-(3-Chlorophenyl)-N'-(2-hydroxy-1,1-dimethylethyl)urea

Supplier: Matrix Scientific

066180-500MG
101931-138EA 243.08 USD
101931-138
N-(3-Chlorophenyl)-N'-(2-hydroxy-1,1-dimethylethyl)urea
N-(3-Chlorophenyl)-N'-(2-hydroxy-1,1-dimethylethyl)urea

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₂
MDL Number: MFCD03488231
CAS Number: 522660-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR