2,3-Dimethylpyrimido[1,2-a]benzimidazol-4(1H)-one

Supplier: Matrix Scientific

066153-500MG
101931-084EA 364.62 USD
101931-084
2,3-Dimethylpyrimido[1,2-a]benzimidazol-4(1H)-one
2,3-Dimethylpyrimido[1,2-a]benzimidazol-4(1H)-one

Hazard: Irritant

Formula: C₁₂H₁₁N₃O
MDL Number: MFCD05703538
CAS Number: 877798-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR