2-Anilino-5-benzyl-6-methylpyrimidin-4(3H)-one

Supplier: Matrix Scientific

066125-500MG
101931-030EA 243.08 USD
101931-030
2-Anilino-5-benzyl-6-methylpyrimidin-4(3H)-one
2-Anilino-5-benzyl-6-methylpyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₁₈H₁₇N₃O
MDL Number: MFCD08723638
CAS Number: 897305-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR