Methyl (5,7-dihydroxy-2-oxo-2H-chromen-4-yl)acetate

Supplier: Matrix Scientific

065072-500MG
101928-944EA 364.62 USD
101928-944
Methyl (5,7-dihydroxy-2-oxo-2H-chromen-4-yl)acetate
Methyl (5,7-dihydroxy-2-oxo-2H-chromen-4-yl)acetate

Hazard: Irritant

Formula: C₁₂H₁₀O₆
MDL Number: MFCD02676912
CAS Number: 809234-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR