2-[(2-Chloro-6-oxo-7,8,9,10-tetrahydro-6H-benzo[c]chromen-3-yl)oxy]propanoic acid

Supplier: Matrix Scientific

065067-500MG
101928-934EA 183.08 USD
101928-934
2-[(2-Chloro-6-oxo-7,8,9,10-tetrahydro-6H-benzo[c]chromen-3-yl)oxy]propanoic acid
2-[(2-Chloro-6-oxo-7,8,9,10-tetrahydro-6H-benzo[c]chromen-3-yl)oxy]propanoic acid

Hazard: Irritant

Formula: C₁₆H₁₅ClO₅
MDL Number: MFCD01832461
CAS Number: 326878-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR