[(2-Oxo-4-phenyl-2H-chromen-7-yl)oxy]acetic acid
Supplier: Matrix Scientific
065052-500MG
101928-904EA
146.15
USD
101928-904
[(2-Oxo-4-phenyl-2H-chromen-7-yl)oxy]acetic acid
[(2-Oxo-4-phenyl-2H-chromen-7-yl)oxy]acetic acid
Hazard: Irritant
Formula:
C₁₇H₁₂O₅ |
MDL Number:
MFCD00708042 CAS Number: 130181-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...