[4-(8-Methoxy-2-oxo-2H-chromen-3-yl)phenoxy]acetic acid

Supplier: Matrix Scientific

065035-500MG
101928-870EA 183.08 USD
101928-870
[4-(8-Methoxy-2-oxo-2H-chromen-3-yl)phenoxy]acetic acid
[4-(8-Methoxy-2-oxo-2H-chromen-3-yl)phenoxy]acetic acid

Hazard: Irritant

Formula: C₁₈H₁₄O₆
MDL Number: MFCD09996812
CAS Number: 1010924-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR