6,7-Dihydroxy-4-propyl-2H-chromen-2-one
Supplier: Matrix Scientific
065009-500MG
101928-818EA
183.08
USD
101928-818
6,7-Dihydroxy-4-propyl-2H-chromen-2-one
6,7-Dihydroxy-4-propyl-2H-chromen-2-one
Hazard: Irritant
Formula:
C₁₂H₁₂O₄ |
MDL Number:
MFCD08741836 CAS Number: 15941-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...