3-Benzyl-6,7-dihydroxy-4-methyl-2H-chromen-2-one

Supplier: Matrix Scientific

064942-500MG
101928-684EA 183.08 USD
101928-684
3-Benzyl-6,7-dihydroxy-4-methyl-2H-chromen-2-one
3-Benzyl-6,7-dihydroxy-4-methyl-2H-chromen-2-one

Hazard: Irritant

Formula: C₁₇H₁₄O₄
MDL Number: MFCD08741838
CAS Number: 23368-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR