[3-(3-Methoxyphenyl)-6-oxopyridazin-1(6H)-yl]acetic acid

Supplier: Matrix Scientific

064936-500MG
101928-672EA 364.62 USD
101928-672
[3-(3-Methoxyphenyl)-6-oxopyridazin-1(6H)-yl]acetic acid
[3-(3-Methoxyphenyl)-6-oxopyridazin-1(6H)-yl]acetic acid

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₄
MDL Number: MFCD16090097
CAS Number: 1206118-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR