[(2-Oxo-3-phenyl-2H-chromen-7-yl)oxy]acetic acid

Supplier: Matrix Scientific

064779-500MG
101928-358EA 243.08 USD
101928-358
[(2-Oxo-3-phenyl-2H-chromen-7-yl)oxy]acetic acid
[(2-Oxo-3-phenyl-2H-chromen-7-yl)oxy]acetic acid

Hazard: Irritant

Formula: C₁₇H₁₂O₅
MDL Number: MFCD09843368
CAS Number: 141113-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR