[2-(3-Thienyl)ethyl]amine

Supplier: Matrix Scientific

Synonyms: 2-(3-Thienyl)ethylamine

064699-500MG
101928-198EA 727.69 USD
101928-198
[2-(3-Thienyl)ethyl]amine
[2-(3-Thienyl)ethyl]amine

Hazard: Irritant

Formula: C₆H₉NS
MW: 127.21 g/mol
MDL Number: MFCD00176329
CAS Number: 59311-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR