6-Chloro-5-fluoropyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-6-chloro-5-fluoropyrimidine

064634-1G
101928-076EA 43.08 USD
101928-076
6-Chloro-5-fluoropyrimidin-4-amine ≥95%
6-Chloro-5-fluoropyrimidin-4-amine

Hazard: Irritant-Harmful/Avoid Skin Contact

Formula: C₄H₃ClFN₃
MW: 147.5400 g/mol
MDL Number: MFCD14706712
CAS Number: 851984-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR