3-Bromo-2-thiophenesulfonyl chloride
Supplier: Matrix Scientific
Synonyms:
3-Bromothiophene-2-sulfonyl chloride
, 3-Bromothiophene-2-sulphonyl chloride
064619-500MG
101928-048EA
606.15
USD
101928-048
3-Bromo-2-thiophenesulfonyl chloride
3-Bromo-2-thiophenesulphonyl chloride
Hazard: Irritant
Formula:
C₄H₂BrClO₂S₂ |
MDL Number:
MFCD08059484 CAS Number: 170727-02-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...