4-Amino-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: H-Phe(4-NH2)-OHH-p-Amino-Phe-OH

064573-500MG
101927-952EA 146.15 USD
101927-952
4-Amino-L-phenylalanine
4-Amino-L-phenylalanine

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MW: 180.207 g/mol
MDL Number: MFCD00069927
CAS Number: 943-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR