4-[(Trimethylsilyl)ethynyl]benzaldehyde

Supplier: Matrix Scientific

Synonyms: 4-((Trimethylsilyl)ethynyl)benzaldehyde

064567-500MG
101927-938EA 243.08 USD
101927-938
4-[(Trimethylsilyl)ethynyl]benzaldehyde
4-[(Trimethylsilyl)ethynyl]benzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₄OSi
MW: 202.33 g/mol
MDL Number: MFCD02093765
CAS Number: 77123-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR