4-Chloro-2-(5-isoxazolyl)phenol

Supplier: Matrix Scientific

064560-500MG
101927-924EA 136.85 USD
101927-924
4-Chloro-2-(5-isoxazolyl)phenol
4-Chloro-2-(5-isoxazolyl)phenol

Hazard: Irritant

Formula: C₉H₆ClNO₂
MDL Number: MFCD00218660
CAS Number: 86176-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR