2-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid

Supplier: Matrix Scientific

064493-500MG
101927-812EA 323.08 USD
101927-812
2-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid
2-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
CAS Number: 27363-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR