4-Carbamoyltetrahydropyran

Supplier: Matrix Scientific

Synonyms: Tetrahydro-2H-pyran-4-carboxamide

064403-500MG
101927-630EA 484.62 USD
101927-630
4-Carbamoyltetrahydropyran
4-Carbamoyltetrahydropyran

Hazard: Irritant

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD08235066
CAS Number: 344329-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR