4,6-Dihydroxy-2-methylpyrimidine

Supplier: Matrix Scientific

Synonyms: 2-Methylpyrimidine-4,6-diol2-Methyl-4,6-pyrimidinediol

064335-500MG
101927-498EA 243.08 USD
101927-498
4,6-Dihydroxy-2-methylpyrimidine
4,6-Dihydroxy-2-methylpyrimidine

Hazard: Irritant

Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
MDL Number: MFCD00006104
CAS Number: 1194-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR