4'-Methoxy-3'-nitroacetophenone

Supplier: Matrix Scientific

064226-500MG
101927-304EA 181.7 USD
101927-304
4'-Methoxy-3'-nitroacetophenone
4'-Methoxy-3'-nitroacetophenone

Hazard: Irritant

Formula: C₉H₉NO₄
MDL Number: MFCD00017023
CAS Number: 6277-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR