7-(Trifluoromethyl)isatin
Supplier: Matrix Scientific
064157-500MG
101927-164EA
183.08
USD
101927-164
7-(Trifluoromethyl)isatin
7-(Trifluoromethyl)isatin
Hazard: Irritant
Formula:
C₉H₄F₃NO₂ MW: 215.1300 g/mol |
MDL Number:
MFCD01248408 CAS Number: 391-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...