6-Bromo-1H-[1,2,3]triazolo[4,5-b]pyridine

Supplier: Matrix Scientific

062811-500MG
101924-596EA 364.62 USD
101924-596
6-Bromo-1H-[1,2,3]triazolo[4,5-b]pyridine
6-Bromo-1H-[1,2,3]triazolo[4,5-b]pyridine

Hazard: Irritant

Formula: C₅H₃BrN₄
MDL Number: MFCD04060716
CAS Number: 92276-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR