Methyl-3-amino-1H-pyrazole-4-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-1H-pyrazole-4-carboxylate

062807-500MG
101924-588EA 243.08 USD
101924-588
Methyl-3-amino-1H-pyrazole-4-carboxylate
Methyl-3-amino-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₅H₇N₃O₂
MW: 141.1300 g/mol
MDL Number: MFCD02936636
CAS Number: 29097-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR