(4-Ethyl-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid
Supplier: Matrix Scientific
062767-500MG
101924-522EA
362.25
USD
101924-522
(4-Ethyl-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid
(4-Ethyl-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid
Hazard: Irritant
Formula:
C₉H₁₄N₂O₂ |
MDL Number:
MFCD12030897 CAS Number: 1177352-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...