Fluorene-4-carboxylic acid
Supplier: Matrix Scientific
062631-500MG
101924-254EA
364.62
USD
101924-254
Fluorene-4-carboxylic acid
Fluorene-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₀O₂ |
MDL Number:
MFCD00001112 CAS Number: 6954-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...